Print…
Thermometer clamp, with swivelling bosshead
Fornecedor: BOCHEM
5621
BOCH5621EA 43.1 EUR
BOCH5621
Thermometer clamp, with swivelling bosshead
Pinças /Clamps Pinças de uso geral
These chromium plated clamps are used for holding thermometers (Ø 6 to 15 mm). Swivelling bosshead (Ø 16,5 mm) can be rotated and locked at any angle (0 to 360°).