Print…
Suportes para tubos de ensaio
Fornecedor: BOCHEM
2283
BOCH2283EA 13.1 EUR
BOCH2283
Suportes para tubos de ensaio
Pinças /Tongs
This test tube holder is made from stainless steel (18/10) and features a wooden handle. For tubes with a diameter from 10 to 25 mm.