Print…
Syringe filters, nylon
Fornecedor: Ahlstrom-Munksjö
6012N13002
BINZ6012N13002EA 104 EUR
BINZ6012N13002
Syringe filters, nylon
Filtros Filtros de Seringa
This syringe filter with hydrophilic polyamide membrane is used for sterilisation and clarification of biological fluids, buffer solutions and tissue culture medias. Also suitable for filtration of aqueous and organic solutions in HPLC.
  • Good mechanical strength and strong absorption
  • Resistance against most of organic solvents and most of alkaline liquids
  • Chemical compatibility: pH ranges from 3 to 14
  • Filtration and clarification of solvents