Print…
Right leg skeleton
Fornecedor: 3B Scientific
1000091
BINH1000091EA 0 EUR
BINH1000091
Right leg skeleton
Modelos Biologia humana
This realistic anatomical replica of the human leg skeleton is wire mounted. The leg skeleton is slightly flexible through toes and ankle joint.
  • For increased realism not only the leg skeleton part is flexible, but also is the knee joint.