Print…
Solvent-based paint pen markers, Bel-Art Products, a part of SP
Fornecedor: Bel-Art Products, a Part of SP
F133770000 F133770001
BELAF133770000EA 223 EUR
BELAF133770000 BELAF133770001
Solvent-based paint pen markers, Bel-Art Products, a part of SP
Canetas Washable Markers
Writes on all plastics, glass, metal, wood, paper and cloth, including Teflon® fluoropolymer resins.
  • Precision seal valve provides smooth ink flow and preserves the pen’s life, even if left uncapped
  • When dry, the solvent based ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
  • Markings are insoluble in water or hydrocarbons
  • Resists many solvents, but is easily removed with alcohol or acetone