Print…
Peristaltic pumps, PLP series, behrotest®
Fornecedor: BEHR
B00210217 B00210220
BEHRB00210217EA 802 EUR
BEHRB00210217 BEHRB00210220
Peristaltic pumps, PLP series, behrotest®
Bombas Bombas Peristálticas
Models PLP 66, PLP 330 and PLP 1000. These peristaltic pumps include a pump rotor, two compression and four guide rollers.
  • Ready for immediate use
  • Adjustable flow rate
  • No tools requires to change pump tubing
The behrotest peristaltic pumps PLP are suitable for the discontinued transport of watery solutions / non aggressive applications. They find their application in the laboratory, the food industry as well as in the analytical, medicinal or biological field. The RPM (rotation per minute) can be adjusted manually and the flow direction can beMais informação do produto