Print…
Digital hotplates, Cimarec+™
Fornecedor: Thermo Fisher Scientific
HP88854105 HP88857105 HP88850105
BARNHP88854105EA 282 EUR
BARNHP88854105 BARNHP88857105 BARNHP88850105
Digital hotplates, Cimarec+™
Aquecimentos Placas de aquecimento
These hotplates feature a raised display design which protects the electronics from any liquid spills.
  • Easy-to-read LED display for heating temperature
  • Hot top warning indicator and automatic over-temperature protection
  • Temperature adjustable in 1 degree increments
  • Top surface temperature could rise to the maximum
  • 12 litre capacity
  • Heating up time is 8 min to maximum temperature
Temperature control accuracy (at 37 °C): ≤±5 °C
Temperature uniformity (at 37 °C): ≤±2 °C
Fornecimento: Supplied with cCSAUS certificate. Other accessories such as optionalMais informação do produto