Print…
Deionizer, barnstead Bantam™
Fornecedor: Thermo Fisher Scientific
D0805
BARND0805EA 1560 EUR
BARND0805
Deionizer, barnstead Bantam™
Sistemas para purificação da água Desionizadores
It is a simple, cost effective method for deionisation or pretreatment of lab water. Typically these units are ideal where water usage is up to 38 litres per day.
Encomendar
ESPECIFICAÇÕES
Modelo Bantam™
Environmentally Preferable
Capacidade 38 L/day
W×D×H 220×150×720 mm