Print…
Incubating orbital shakers, MaxQ™ HP
Fornecedor: Thermo Fisher Scientific
238000 238001 238008 238002
BARN238000EA 2270 EUR
BARN238000 BARN238001 BARN238008 BARN238002
Incubating orbital shakers, MaxQ™ HP
Agitadores Incubadoras com agitação
Max Q™ HP combines incubated or refrigerated temperature with an orbital shaker.
  • Triple counter-balanced mechanism
  • Microprocessor functions are calibrated through keypad
  • Clear acrylic lid withstands years of use without cracking or discoloration
  • Audible over and under-temperature alarms ensure sample protection
  • Innovative design ensures system protection in case of spills
Features a double walled insulated chamber which ensures temperature uniformity of ±0,3 °C. The spacious stainless steel chamber allows stacking of a 457×457 mm platform to hold six 2Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Modelo
Movimento de agitação
Environmentally Preferable
Gama de temperaturas
Gama de velocidades
Max. load
Exactidão da temperatura a 37 °C
L×P da plataforma
L×P×A internas
L×P×A
Peso
Shipping weight
Orbit