Print…
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Fornecedor: Corning
FDSS-1536-BK-R
AXYGFDSS-1536-BK-REA 8389 EUR
AXYGFDSS-1536-BK-R
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Pontas de pipetas
These black automation tips made of PP are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Conductive tips
  • Certified DNase-/RNase-free and non pyrogenic for complete protection against contamination
  • Optimised fine point for accurate dispensing