Print…

Anti-HFE Rabbit Polyclonal Antibody

Fornecedor: Aviva Systems Biology
ARP44285_P050
AVIVARP44285_P050EA 376 EUR
AVIVARP44285_P050
Anti-HFE Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: HFE (hemochromatosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR