Print…
Immersion oil
Fornecedor: PanReac AppliChem
A0699.0100
APPCA0699.0100EA 63.6 EUR
APPCA0699.0100
Immersion oil
Corantes Microscopy and staining reagents
This fluorescence-free immersion oil for microscopy is highly UV-permeable and free of PCB's.