Print…

6-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester

Fornecedor: Apollo Scientific

Sinônimos: tert-Butyl 6-aminoindoline-1-carboxylate, tert-Butyl 6-amino-2,3-dihydro-1H-indole-1-carboxylate

OR305255-1G
APOSOR305255-1GEA 306 EUR
APOSOR305255-1G
6-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
6-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester
Fórmula: C₁₃H₁₈N₂O₂
Peso Molecular: 234.3 g/mol
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD08059271
Número CAS: 129488-00-2

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR