Print…

Anti-Nucleolin Rabbit Recombinant Antibody [clone: NCL/7014R]

Fornecedor: ANTIBODIES.COM
A278037-100
ANTIA278037-100EA 520 EUR
ANTIA278037-100
Anti-Nucleolin Rabbit Recombinant Antibody [clone: NCL/7014R]
Anticorpos
Recombinant rabbit monoclonal [NCL/7014R] antibody to Nucleolin for Flow Cytometry, IF, WB and IHC-P with samples derived from human.

  • Validated Applications: Flow Cytometry, IF, WB, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Nucleolin
Clonality: Recombinant
Clone: NCL/7014R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR