Print…

Anti-Nucleolin Goat Polyclonal Antibody

Fornecedor: ANTIBODIES.COM
A121665-100
ANTIA121665-100EA 470 EUR
ANTIA121665-100
Anti-Nucleolin Goat Polyclonal Antibody
Anticorpos
Goat polyclonal antibody to Nucleolin for WB with samples derived from human, rat, mouse, canine and monkey.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1,000 to 1:5,000

Type: Primary
Antigen: Nucleolin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Mouse, Dog, Monkey
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR