Print…

Anti-PIST Rabbit Monoclonal Antibody

Fornecedor: Abcam
AB133472-100
ABCAAB133472-100EA 1030 EUR
ABCAAB133472-100
Anti-PIST Rabbit Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: PIST
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR