Print…

hBSA *Optimized for Maximum Immunization Response* (10 mg)

Fornecedor: AAT BIOQUEST
5601
AATB5601EA 307 EUR
AATB5601
hBSA *Optimized for Maximum Immunization Response* (10 mg)
Soro
hBSA is the bovine serum albumin modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).

  • Lyophilized powder
  • Excellent quality control
  • Stronger immune response
  • 60000 molecular weight

Atenção: For research use only.
Encomendar

ESPECIFICAÇÕES

Environmentally Preferable
Purification 97%, via fractionation
Temperatura de armazenamento <–20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR