Print…

Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin)/iFluor® 647)

Fornecedor: AAT BIOQUEST
50240 50241
AATB50240EA 220 EUR
AATB50240 AATB50241
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin)/iFluor® 647)
Anticorpos
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Atenção: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/iFluor® 647
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR