Print…

Anti-IgG Goat Polyclonal Antibody (XFD700)

Fornecedor: AAT BIOQUEST
50176 50177
AATB50176EA 120 EUR
AATB50176 AATB50177
Anti-IgG Goat Polyclonal Antibody (XFD700)
Anticorpos
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Atenção: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: XFD700
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Novo produto
Encomendar

ESPECIFICAÇÕES

Símbolo do antígeno IgG
Nome do antigénio Immunoglobulin G
Environmentally Preferable
Tipo de anticorpo Secundário
Clonalidade Policlonal
Conjugado XFD700
Reatividade Human
Hospedeiro Goat
Isótipo IgG (H, L)
Western blot Yes
ELISA Yes
ImunoQuímica Yes
Citometria de fluxo Yes
Adsorção cruzada No
Temperatura de armazenamento Freeze (<–15 °C); Minimise light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR