Print…
ATTO 647N acid
Fornecedor: AAT BIOQUEST
2854
AATB2854EA 775 EUR
AATB2854
ATTO 647N acid
ATTO 647N acid
ATTO 647N is a rhodamine-based fluorescent dye optimized for use in the red spectral region, with similar spectral characteristics as Cy5.
  • Stable for 12 months
Quality tested in-house
Atenção: For Research Use Only