Print…
2’,3’-cGAMP-iFluor 488 conjugado
Fornecedor: AAT BIOQUEST
20320
AATB20320EA 1330 EUR
AATB20320
2’,3’-cGAMP-iFluor 488 conjugado
2’,3’-cGAMP-iFluor 488
iFluor® 488-labeled cGAMP provides an excellent fluorescent probe for developing fluorescence-based assays and tests for detecting cGAMP or monitoring its activity.
  • Stable for 12 months
Quality tested in-house
Atenção: For Research Use Only