Print…
3'-O-Azidomethyl-dGTP
Fornecedor: AAT BIOQUEST
17224 17225
AATB17224EA 673 EUR
AATB17224 AATB17225
3'-O-Azidomethyl-dGTP
Reagentes de ácidos nucleicos Nucleótidos
3'-O-Azidomethyl-dGTP is one of the four essential dNTPs (A, C, G, T) that can be used for the DNA sequencing by synthesis (SBS).
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Atenção: For research use only.