Print…
Buccutite™ Rapid APC Antibody Labelling Kits (2 Labellings)
Fornecedor: AAT BIOQUEST
1313 1311
AATB1313EA 460 EUR
AATB1313 AATB1311
Buccutite™ Rapid APC Antibody Labelling Kits (2 Labellings)
Sistemas de ligação e rotulagem de proteínas
APC is an red fluorescent protein which has an excitation wavelength of 651 nm and an emission wavelength of 662 nm.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol