Print…

ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

Fornecedor: PanReac AppliChem

Sinônimos: NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)

A1395.0100 A1395.0500 A1395.0001
A1395.0100EA 125 EUR
A1395.0100 A1395.0500 A1395.0001
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Fórmula: C₂₁H₂₆N₇Na₄O₁₇P₃
Peso Molecular: 833.35 g/mol
Número MDL : MFCD00036263
Número CAS: 2646-71-1
EINECS: 220-163-3

Encomendar

ESPECIFICAÇÕES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR