Print…

Metanol ≥99.9%, BAKER ANALYZED® LC-MS para HPLC, para espectrofotometria UV, J.T.Baker®

Fornecedor: Avantor
Perigo

Sinônimos: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Management of Change (MOC) category = HR

9830-03EA 222.2 EUR
9830-03 9830-02
Metanol ≥99.9%, BAKER ANALYZED® LC-MS para HPLC, para espectrofotometria UV, J.T.Baker®
Metanol

Pure, trusted chromatography solvents providing high selectivity, reproducibility and accuracy of results that come in a variety of grades such as HPLC, LCMS, ULTRA LCMS and PHOTREX.

Fórmula: CH₃OH
Peso Molecular: 32.04 g/mol
Ponto de ebulição: 64.6
Ponto de Fusão: –98 °C
Densidade: 0.7918 g/cm³ (20 °C)
Ponto de Inflamação: 11 °C (rec. fechado)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00004595
Número CAS: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
ALCANCE: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

For Laboratory,Research,or Manufacturing Use = 99.9 %
Assay (CH₃OH) (by GC, corrected for water) ≥ 99.9 %
Appearance Passes Test
Gradient Elution Test (a.u.) - 254 nm ≤ 0.01
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤ 0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 1.0 ppb
Color (APHA) ≤ 10
Residue after Evaporation ≤ 1.0 ppm
Water (H₂O) ≤ 500 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤ 50 ppb
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Trace Impurities - * Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lithium (Li) ≤ 30.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Nickel (Ni) ≤ 30.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - * Sodium (Na) ≤ 50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR