Print…
Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
Fornecedor: Merck
96442-100ML
96442-100MLEA 185 EUR
96442-100ML
Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
Padrões Cationic Standards Multi-Element Standards
100 mg/l each: Ti, Zr, Hf, Nb, Ta, Mo, W and Re in 2% nitric acid and 1% hydrofluoric acid
Certificações: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034