Print…
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Fornecedor: Merck
91286-100ML
91286-100MLEA 219 EUR
91286-100ML
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Padrões Cationic Standards Multi-Element Standards
Ca, 1000 mg/l
K, 200 mg/l
Li, 50 mg/l
Mg, 200 mg/l
Na, 200 mg/l
NH4, 400 mg/l
Certificações: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034