Print…
VWR®LABWASH® Premium USB, Detergents and cleaning agents for ultrasonic baths
Fornecedor: VWR Chemicals
88287.290EA 3.65 EUR
88287.290 88287.360 88287.440 88288.290 88288.360 88288.440
VWR®LABWASH® Premium USB, Detergents and cleaning agents for ultrasonic baths
Soluções de limpeza Detergentes e Limpadores de Superfícies
A range of cleaning agents designed for use in ultrasonic baths.
VWR® LABWASH® Premium USB 13 is an intensive and thorough product for removing organic deposits such as oils, resins, greases from equipment and tools; especially in ultrasonic baths.
The product contains special corrosion inhibitors to increase material compatibility. Contained dispersants in combination with surfactants provide fast and intensive cleaning.
Working concentration: 2 - 15% (without ultrasonic); 0,5 - 8% (with ultrasonic).
VWR® LABWASH® Premium USB 33 Universal is specially designed for cleaning inMais informação do produto