Print…
VWR® PESTINORM®, Dichloromethane (DCM) Stabilised, Secondary Reference Standard for GC
Fornecedor: VWR Chemicals
85658.180EA 182 EUR
85658.180
VWR® PESTINORM®, Dichloromethane (DCM) Stabilised, Secondary Reference Standard for GC
Padrões Padrões de Cromatografia Gasosa
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID
Stabilised with amylene.