Print…

Phytochemical standard, Fraxin, analytical standard, Supelco®

Fornecedor: Merck

80533-10MG
80533-10MGEA 288 EUR
80533-10MG
Phytochemical standard, Fraxin, analytical standard, Supelco®
Padrões Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8ac3b87c22ec815baf75a69f51e1785a40a672f&vid=e8ac3b87c22ec815baf75a69f51e1785a40a672f&prid=web&clid=SAPDAM

Encomendar

ESPECIFICAÇÕES

Descrição Fraxin
Environmentally Preferable
CAS No. 524-30-1
Formula C₁₆H₁₈O₁₀

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR