Print…
Immobilised Pepsin
Fornecedor: G Biosciences
786-791
786-791EA 519 EUR
786-791
Immobilised Pepsin
Extracção de proteínas Reagentes de Extração de Proteínas
A proteolytic enzyme immobilised on 4% agarose that is routinely used for the generation of F(ab’)₂ fragments from Immunoglobulin G (IgG). The pepsin has the ability to cleave the heavy chains near the hinge region. The immobilised Pepsin offers the distinct advantage of eliminating enzyme contamination of the F(ab)₂ fragments.
  • Generate F(ab)₂ fragments
  • Eliminate contaminating pepsin enzyme
  • Can be used in virtually all scenarios using free pepsin