Print…
Endotoxins and pyrogens removal reagent, EndotoxinOUT™
Fornecedor: G Biosciences
786-369 786-368 786-367
786-369EA 377 EUR
786-369 786-368 786-367
Endotoxins and pyrogens removal reagent, EndotoxinOUT™
Soluções de limpeza Descontaminantes
EndotoxinOUT™ consists of 6% cross-linked agarose covalently linked to polymyxin B to bind and remove harmful pyrogens from a solution.
Polymyxin B is a family, polymyxin B1 and B2, of antibiotics that bind to the negatively charged site of the lipid A portion of bacterial lipopolysaccharide layer neutralizing the endotoxic activity. The covalent coupled agarose and polymyxin B is a stable matrix that resists leaching. An ideal product for the clean up of buffers, cell culture media, protein solutions, nucleic acid (DNA) samples and pharmacological components. The fractionation range is 10 to 4000 kDa.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable