Print…
Proteome kits, FOCUS™ Insect Proteome
Fornecedor: G Biosciences
786-360
786-360EA 652 EUR
786-360
Proteome kits, FOCUS™ Insect Proteome
Extracção de proteínas Protein Extraction Kits
Extracts and solubilises nearly all of the proteins from insect cell cultures (i.e. Sf9 and Sf21), including membrane as well as soluble proteins, using a strong chaotropic extraction buffer to solubilise even the most difficult proteins.
  • Single step extraction protocol
  • Supplied with a strong proprietary chaotropic extraction buffer
  • For the extraction of protein from 50×50 μl insect cell pellets
  • Suitable for sample preparation for 2D gel electrophoresis and other applications