Print…

Cloreto de guanidínio 8 mol/l aqueous solution

Fornecedor: G Biosciences
Atenção

Sinônimos: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

786-279
786-279EA 618 EUR
786-279
Cloreto de guanidínio 8 mol/l aqueous solution
Cloreto de guanidínio

A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.


Available as a powder or 8M solution

Fórmula: NH₂C(=NH)NH₂·HCl
Peso Molecular: 95.53 g/mol
Ponto de Fusão: 185
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00013026
Número CAS: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR