Print…

Hidrogenocarbonato de sódio 99.0-100.5% (base seca), pó, GenAR® USP para biotecnologia, Macron Fine Chemicals™

Fornecedor: Avantor Performance Materials

Sinônimos: Bicarbonato de sódio

Management of Change (MOC) category = HR

7749-24EA 764.5 EUR
7749-24 7749-88
Hidrogenocarbonato de sódio 99.0-100.5% (base seca), pó, GenAR® USP para biotecnologia, Macron Fine Chemicals™
Hidrogenocarbonato de sódio
Fórmula: NaHCO₃
Peso Molecular: 84.01 g/mol
Ponto de Fusão: 270
Densidade: 2.159 g/cm³ (20 °C)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00003528
Número CAS: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Food GMP Manufactured Product
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Limit of Ammonia ≤ 20 ppm
USP - Limit of Sulfur Compounds ≤ 0.015 %
USP - Arsenic (As) ≤ 2 ppm
USP - Normal Carbonate Passes Test
USP - Chloride (Cl) ≤ 0.015 %
USP - Identification A (Sodium) Passes Test
USP - Identification B (Bicarbonate) Passes Test
USP - Insoluble Substances Passes Test
USP - Loss on Drying ≤ 0.25 %
EP/BP - Assay (NaHCO₃) 99.0 - 101.0 %
EP/BP - Ammonium (NH₄) ≤ 20 ppm
EP/BP - Appearance of Solution Passes Test
EP/BP - Calcium (Ca) ≤ 100 ppm
EP/BP - Carbonate (pH) ≤ 8.6
EP/BP - Chloride (Cl) ≤ 150 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Iron (Fe) ≤ 20 ppm
EP/BP - Sulfate (SO₄) ≤ 150 ppm
JP - Assay (NaHCO₃) 99.0 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 7.9 - 8.4
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.040 %
JP - Carbonate (CO₃) Passes Test
JP - Ammonium (NH₄) Passes Test
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Arsenic (As) ≤ 2 ppm
FCC - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Limit of Ammonia (NH₃) Passes Test
Endotoxin Concentration (EU/g) ≤ 5.0
No Class 1,2,3 or other solvents are used or produced in the ≤ 5.0
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR