Print…
VWR®, Confocal Dishes for Microscopy
Fornecedor: VWR Collection
734-2904EA 110 EUR
734-2904 734-2906
VWR®, Confocal Dishes for Microscopy
Placas para cultura Placas para Cultura de Células
Confocal dishes are suitable for fluorescent, confocal and phase-contrast microscopy experiments. These dishes combine the convenience of standard 35 mm, PS cell culture dishes with the optical benefits of glass. Dishes are available with Ø 20 mm glass centre, with or without a tissue culture-treated surface.
  • Made from homogenous borosilicate glass with low bubble and inclusion content
  • Transparent, medical grade glue, achromatic
  • Dish surface is smooth and free from striations to maximise usable area for growth
  • Rim on upper side of the lid mates with the