Print…
VWR® Bottle Assemblies, PETG, Sterile, Single-Use
Fornecedor: VWR Collection
734-2844EA 940 EUR
734-2844 734-2845 734-2846 734-2850 734-2851 734-2894
VWR® Bottle Assemblies, PETG, Sterile, Single-Use
Frascos Frascos para meios
Sterile PETG containers, with weldable tubings, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They come in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶