Print…
Cell culture flasks, Nunc™
Fornecedor: Thermo Fisher Scientific
153732 156340 156367 156472 156499 156502 156800 159910 159920 163371 178883 178905 169900 159926 178983 159933 132707 132708 132703 132704 132705 132706 178885 178985 156505 159934 132097 132098
734-2046EA 108 EUR
734-2046 734-2063 734-2064 734-2065 734-2066 734-2067 734-2069 734-2167 734-2168 734-2081 734-2129 734-2131 734-2104 734-1190 734-1340 734-1337 734-2285 734-2286 734-2281 734-2282 734-2283 734-2284 392-0390 392-0391 392-0389 734-1338 734-2608 734-2609
Cell culture flasks, Nunc™
Balões Frascos de Cultura de Células
These Nunc™ cell culture flasks are designed for culture consistency, cell health, and reproducibility. Choose from a variety of surfaces and sizes with culture areas ranging from 25 to 500 cm² to suit your specific applications and cell types.
  • Nunclon Delta certification including testing on four different cell types is provided in each case
  • Barcoded 175 cm² flasks (*) with large Code 128 are compatible with automation
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Flasks treated with the proprietary Nunclon™ Delta surfaceMais informação do produto