Print…
Inserts para cultura de células Falcon®
Fornecedor: Corning
353091 353092 353090 353093 353095 353096 353097 353102 353103 353104 353180 353181 353182 353292 353492 353493 353494 353495
734-0033EA 457.21 EUR
734-0033 734-0034 734-0032 734-0035 734-0036 734-0037 734-0038 734-0040 734-0041 734-0042 734-0051 734-0052 734-0053 734-0060 734-0397 734-0061 734-0062 734-0063
Inserts para cultura de células Falcon®
Microplacas Insertos e Placas
Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture