Print…
Mini tube system, Axygen®
Fornecedor: Corning
MTS-11-C MTS-11-8-C-R-S MTS-06-C-R MTS-06-C-R-S MTS-06-C MTS-11-12-C MTS-11-12-C-R MTS-11-12-C-RS MTS-11-8-C MTS-11-8-C-R MTS-11-C-R MTS-11-C-R-S
733-1979EA 34.07 EUR
733-1979 732-1138 731-0211 731-0212 731-0210 731-0213 731-0214 731-0215 731-0216 731-0217 731-0219 731-0220
Mini tube system, Axygen®
Tubos Microtubos
PP, 100% virgin. The Axygen 96-well mini tube system is designed to work with 8-strip or 12-strip plug caps.
  • Compatible with Beckman, Tecan, Zymark and other robotic workstations
  • One-way lid design with etched in alphnumeric grid ensures positive sample identification
  • Moulded in condensation rings on cover minimise risk of cross-contamination