Print…
Dialysers, D-Tube™ and D-Tube96™
Fornecedor: Merck Millipore (Novagen)
71505-3 71507-3 71504-3 71506-3 71510-3 71508-3 71509-3 71712-3 71713-3 71739-3 71739-4 71740-3 71740-4 71742-3 71742-4 71743-3 71743-4 71746-3 71746-4 71745-3 71745-4
71505-3EA 103 EUR
71505-3 71507-3 71504-3 71506-3 71510-3 71508-3 71509-3 71712-3 71713-3 71739-3 71739-4 71740-3 71740-4 71742-3 71742-4 71743-3 71743-4 71746-3 71746-4 71745-3 71745-4
Dialysers, D-Tube™ and D-Tube96™
Tubos Tubos para diálise
D-Tube96™ Dialysers allow convenient, high throughput dialysis of 96 samples simultaneously. The device features the advantages of the D-Tube™ Dialyzer Mini in a 96-tube format. D-Tube Dialysers are easy to handle tubes with dual membranes providing a large surface area for fast, efficient dialysis. The membrane is ultra clean, EDTA-treated, regenerated cellulose that is sulfur and heavy metal-free.
  • Dual membrane surface for efficient dialysis
  • Convenient to use with single or multichannel pipette or robotic dispensing