Print…
Protein extraction reagents, CytoBuster™
Fornecedor: Merck Millipore (Novagen)
71009-3 71009-4
71009-3EA 145 EUR
71009-3 71009-4
Protein extraction reagents, CytoBuster™
Extracção de proteínas Reagentes de Extração de Proteínas
The CytoBuster™ protein extraction reagent is a proprietary formulation of detergents optimised for efficient extraction of soluble proteins from mammalian and insect cells. The gentle, non-ionic composition of the reagent enables isolation of functionally active endogenous or expressed proteins without a need for secondary treatment such as sonication or freeze/thaw.
CytoBuster™ protein extraction reagent has been specifically formulated for use in Western blotting, immunoprecipitation, and kinase/phosphatase assays. The reagent is compatible with protease inhibitors, kinase inhibitors, and phosphatase inhibitors.