Print…
Swabs with large circular foam head, CleanFoam®
Fornecedor: Texwipe
TX708A
710-0548EA 359 EUR
710-0548
Swabs with large circular foam head, CleanFoam®
Zaragatoas Zaragatoas para limpeza
Thermally-bonded swabs with 100 pore per inch porosity.
  • Head will not crack or chip
  • Inherent softness and low abrasion protect delicate components
  • Large circular head on a long and easy grip handle
Can be used in industrial applications, from general assembly to cleanrooms, requiring high absorbency with low particle generation and uniform dispersing of solvents.
Embalagem: Cleanroom processed bag-within-a-bag.
Atenção: * O (s) produto (s) nesta página não se enquadra no âmbito da Diretiva da UE 2019/904