Print…
Temperature measuring for urine
Fornecedor: SURESCREEN
TEMP
700-9660EA 50.8 EUR
700-9660
Temperature measuring for urine
Testes para detecção de drogas
For use with the standard collection cup, and used to show if a urine sample is within range of body temperature.
  • Samples outside this temperature range could have been adulterated