Print…
VWR® kuroGEL, Semi-dry blotter
Fornecedor: VWR Collection
700-0416EA 1870 EUR
700-0416
VWR® kuroGEL, Semi-dry blotter
Aparelhos de Blotting
The kuroGEL semi-dry blotter provides great flexibility in a laboratory since it can be used for all types of blotting: Western, Southern and Northern. Blotting is fast and easy, and can typically be completed in 15 to 30 min. Set-up procedures are easy and economical with little buffer needed. The screw down lid adjusts to varying gel thicknesses and sizes, while the platinum coated electrodes provide uniform pressure ensuring even transfers.
  • Suitable for Western, Southern and Northern blots
  • Rapid transfer times
  • Uniform heat dispersion
  • Variable gel
Mais informação do produto