Print…
pET-30a(+) DNA, Novagen®
Fornecedor: Merck Millipore (Novagen)
69909-3
69909-3EA 352 EUR
69909-3
pET-30a(+) DNA, Novagen®
Vectors, Plasmids and Libraries
The pET-30a-c(+) vectors carry an N-terminal His Tag®/thrombin/S Tag™/enterokinase configuration plus an optional C-terminal His tag sequence.
Fornecimento: The pET Vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).