Print…
pH tester, Eutech™ Expert pH
Fornecedor: EUTECH
EXPERTPH
663-0308EA 104 EUR
663-0308
pH tester, Eutech™ Expert pH
Medidores electroquímicos Medidores de pH e multi-paramétricos
This waterproof pocket pH tester is ideal for quick pH measurements in hydroponic gardening, aquaculture, agriculture, pools, education, cooling towers, environmental monitoring, water treatment also food and beverages.
  • Easy to read 4-digit dual-line display with battery life, ready and calibration indicators
  • Simultaneous display of pH and temperature
  • Automatic temperature compensation; automatic buffer recognition and calibration
  • Multi-language operational instructions
  • Upgraded key pad with intuitive interface
  • Symmetrical, leak resistant cap with a fill-
Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Modelo Eutech™ Expert pH
Environmentally Preferable
pH - gama 0,0 - 14,0
pH - resolução 0,1
pH - exactidão ±0,1
pH - calibração Up to 3 points
Gama de temperaturas 0...+50,0 °C
Temperature resolution 0,1 °C
Temperature accuracy ±0,5 °C + 1 least significant digit
Compensação da temperatura Automatic
Alimentação de energia Four batteries (A76 or LR44)
Conformidade IP 67
L×P×A 170×45×30 mm