Print…
Elétrodos Redox, InLab® Redox
Fornecedor: Mettler - Toledo
51343200 51343201 51343202 51343203 51343204 51343205
662-2877EA 292 EUR
662-2877 662-2878 662-2879 662-2880 662-2881 662-2882
Elétrodos Redox, InLab® Redox
Eléctrodos Redox Electrodes
Eletrodos redox com conector S7.
  • Ideal for testing the oxidation/reduction in factory and industrial waste (chromium and cyanide concentration), fish farming and aquariums (water quality), and swimming pools (sanitation levels)
  • ARGENTHAL™ reference system with silver-ion trap (InLab® Redox Micro: without silver-ion trap)
  • EQUITHAL® technology with symmetrical conducting layers guarantees shortest response times despite large changes in temperature
  • Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste
Mais informação do produto