Print…
Orion™ Pro Star MP218 pH/Ion/Conductivity/DO Bench Meters
Fornecedor: Thermo Orion
PSTAR2187 PSTAR2180 PSTAR2185 PSTAR2186
662-2534EA 2390 EUR
662-2534 662-2531 662-2532 662-2533
Orion™ Pro Star MP218 pH/Ion/Conductivity/DO Bench Meters
Medidores electroquímicos Medidores de pH e multi-paramétricos
The Thermo Scientific™ Orion™ Pro Star MP218 pH/ion, conductivity, and dissolved oxygen bench meter is designed to provide accurate, reliable electrochemistry testing. Featuring an intuitive interface, enhanced data reporting and robust functionality, the Orion™ Pro Star MP218 meter delivers advanced performance in a modern and simplified package.
  • Large 7" high-resolution colour display
  • Responsive capacitive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter
Mais informação do produto