Print…
Orion™ Pro Star PH211 pH/ORP Bench Meters
Fornecedor: Thermo Orion
PSTAR2115 PSTAR2117 PSTAR2118 PSTAR2114 PSTAR2116 PSTAR2119 PSTAR2110
662-2501EA 979 EUR
662-2501 662-2503 662-2504 662-2500 662-2502 662-2505 662-2499
Orion™ Pro Star PH211 pH/ORP Bench Meters
Medidores electroquímicos Medidores de pH e multi-paramétricos
The Thermo Scientific™ Orion™ Pro Star PH211 pH bench meter is designed to provide accurate, reliable electrochemistry testing. Featuring an intuitive interface, enhanced data reporting and robust functionality, the Orion™ Pro Star PH211 meter delivers advanced performance in a modern and simplified package.
  • Large 7" high-resolution colour display
  • Responsive capacitive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps optimise bench space
  • Supports
Mais informação do produto